ContactUsLinkComponent
Packed Column Molesieve 5A 80/100 2m 2mm ID Silco Shimadzu 17A/2010 1 * 1 items
Catalog # REST80429-853
Supplier:  Restek
Packed Column Molesieve 5A 80/100 2m 2mm ID Silco Shimadzu 17A/2010 1 * 1 items
Catalog # REST80429-853
Supplier:  Restek
Frequently Bought Together