ContactUsLinkComponent
Packed Column 0.2% Carbowax 1500 80/100 CBK C 2m, 1/8 OD, 2mmID, Silco For HP 6850 GCs, OnCol, Preconditioned 1 * 1 items
Catalog # REST80122-811900
Supplier:  Restek
Packed Column 0.2% Carbowax 1500 80/100 CBK C 2m, 1/8 OD, 2mmID, Silco For HP 6850 GCs, OnCol, Preconditioned 1 * 1 items
Catalog # REST80122-811900
Supplier:  Restek
Frequently Bought Together