1250 pieces for 2500 cell countsJuLI Br single-use cell counting slides for twocell counts each. Including 37.5 ml Trypan Bluesolution (0.4 %).Shipment: RTStorage: RT 1 * 1.250
Catalog # NANOJUBRCC-03
Supplier: NANOENTEK
Frequently Bought Together