ContactUsLinkComponent
THERMO-MAT 384 white, 5 x 5 mats Up to 50 times autoclavable and reusable silicone mat for 384 well plates. Temperature range: -20 C up to +120 C. Effective and uniform sealing when using in thermocyclers with high pressures lid. 1 * 25 items
Catalog # PEQL82-0753-B
Supplier:  VWR Collection
THERMO-MAT 384 white, 5 x 5 mats Up to 50 times autoclavable and reusable silicone mat for 384 well plates. Temperature range: -20 C up to +120 C. Effective and uniform sealing when using in thermocyclers with high pressures lid. 1 * 25 items
Catalog # PEQL82-0753-B
Supplier:  VWR Collection
Frequently Bought Together