ContactUsLinkComponent
PEQSTAR 96Q REAL-TIME CYCLER 4 channel system 4-channel Real-Time PCR cycler with 96-well block format, long-life LED excitation light source, and high-sensitivity photomultiplier detector. Channels: F1: ex470nm/em525nm, FAM, SYBR Green F2: ex523nm/e 1 * 1 items
Catalog # 732-2774
Supplier:  VWR Peqlab
undefined
PEQSTAR 96Q REAL-TIME CYCLER 4 channel system 4-channel Real-Time PCR cycler with 96-well block format, long-life LED excitation light source, and high-sensitivity photomultiplier detector. Channels: F1: ex470nm/em525nm, FAM, SYBR Green F2: ex523nm/e 1 * 1 items
Catalog # 732-2774
Supplier:  VWR Peqlab
Frequently Bought Together