ContactUsLinkComponent
ORBITAL SHAKER PSU-10I Compact orbital shaker Orbital rotation motion Platfrom size 220 x 220 mm Speed range 30 - 450 rpm Orbit 10 mm Digital Timer 1 min - 96 h Max. Loading 3.0 kg Warranty 2 Years 1 * 1 items
Catalog # BSANPSU-10I/UP-12
Supplier:  BIOSAN
undefined
ORBITAL SHAKER PSU-10I Compact orbital shaker Orbital rotation motion Platfrom size 220 x 220 mm Speed range 30 - 450 rpm Orbit 10 mm Digital Timer 1 min - 96 h Max. Loading 3.0 kg Warranty 2 Years 1 * 1 items
Catalog # BSANPSU-10I/UP-12
Supplier:  BIOSAN
Frequently Bought Together