ContactUsLinkComponent
48 WELL PCR PLATE (6 X 8 WELLS) colourless, thin-walled, 1 x 50 plates Semi-skirted plate with 48 wells for medium sample throughput. Autoclavable. Temperature range: -80 C and +121 C. Free of RNases, DNases and human DNA. 1 * 50 items
  732-3219
undefined
48 WELL PCR PLATE (6 X 8 WELLS) colourless, thin-walled, 1 x 50 plates Semi-skirted plate with 48 wells for medium sample throughput. Autoclavable. Temperature range: -80 C and +121 C. Free of RNases, DNases and human DNA. 1 * 50 items
  732-3219
Frequently Bought Together