EUROGOLD PLASMID MINIPREP KIT I EUROCLONE 200 Präparationen Plasmid-Extraktion aus bis zu 10 ml Kultur Ertrag: 15 - 25 µg Kitbestandteile: PerfectBind DNA Columns, Collection Tubes, Solution I, Solution II, Solution III, HB Buffer, DNA Wash Buffer, 1 * 200 Tests
Catalog # PEQLEMR500200
Supplier: VWR Collection
Related Information
Frequently Bought Together