EUROGOLD BLOOD RNA KIT (S-LINE) 5 Präparationen RNA-Extraktionen aus bis zu 1 ml Vollblut. Ertrag: bis 5 µg Kitbestandteile: PerfectBind RNA Columns (Safety-Line), DNA Removing Columns, Collection Tubes, Lysis Buffer ER, RNA Lysis Buffer T, RNA Was 1 * 5 Tests
Catalog # PEQLEMR511005
Supplier: VWR Collection
Frequently Bought Together