PEQGOLD BLOOD DNA MINI KIT PLUS (C-LINE) 5 Preps Isolation of DNA from up to 400 µl whole blood. Yield: up to 30 µg Kit Components: PerfectBind DNA Columns (Classic-Line), Collection Tubes, Binding Solution BL, DNA Lysis Buffer BL, DNA Wash Buffer BL 1 * 5 Tests
Catalog # 732-3054
Supplier: VWR Peqlab
Frequently Bought Together