EUROGOLD GEL EXTRACTION KIT EUROCLONE 5 Prparationen 50-bp- bis 40-kbp-DNA aus Agarosegelen oder Reaktionsanstzen Kitbestandteile: Perfect Bind DNA Columns (Classic Line), Collection Tubes, XP2 Binding Buffer, SPW Wash Buffer, Elution Buffer Vers 1 * 5 Tests
Catalog # PEQLEMR501005
Supplier: VWR Peqlab
Frequently Bought Together