ContactUsLinkComponent
TISAB II (STANDARD METHODS/AOAC) for samples containing <3 ppm in Fe and/or Al 1 * 1.000 mL
Catalog # APPC274765.1211
Supplier:  PanReac AppliChem
TISAB II (STANDARD METHODS/AOAC) for samples containing <3 ppm in Fe and/or Al 1 * 1.000 mL
Catalog # APPC274765.1211
Supplier:  PanReac AppliChem
Related Information
Frequently Bought Together