Orders LinkContactUsLinkComponent
Storage Sacks
Storage Sacks
Approx. 357 bags/pack, 0,06 mm, 400×600 mm
Catalog # LAMA123
Supplier:   LAMAPLAST
Storage Sacks
Catalog # LAMA123
Supplier:  LAMAPLAST
Storage Sacks
Storage Sacks
Approx. 357 bags/pack, 0,06 mm, 400×600 mm
Price
244,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
LDPE, neutral storage bags.
  • With bottom weld
  • Can be used for foodstuffs
Specifications
  • Description:
    Approx. 357 bags/pack
  • Dimensions:
    400×600 mm
  • Seq:
    06
  • Thickness:
    0,06 mm