Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
DENSITY STANDARD 2.0812G/ML @ 20C 1 * 100 mL
REGEDEN20240PY
:
REAGECON
DENSITY STANDARD 2.0812G/ML @ 20C 1 * 100 mL
REGEDEN20240PY
:
REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
: VWR Chemicals
DENSITY STANDARD 1.5799G/ML @ 20C 1 * 100 mL
: REAGECON