ContactUsLinkComponent
Taq Plus DNA Polymerase is a mixture of Taq DNA Polymerase and Pfu, a proofreading DNA Polymerase, which allows for the amplificatio n of long templates, up to 20kb, with high fidelity. 1 * 40 Tests
  GENO786-851
Taq Plus DNA Polymerase is a mixture of Taq DNA Polymerase and Pfu, a proofreading DNA Polymerase, which allows for the amplificatio n of long templates, up to 20kb, with high fidelity. 1 * 40 Tests
  GENO786-851
Frequently Bought Together