ContactUsLinkComponent
BALANCE ME204 ANALYTICAL 220G X 0.1MG 1 * 1 items
Catalog # METB30029066
Supplier:  Mettler - Toledo
BALANCE ME204 ANALYTICAL 220G X 0.1MG 1 * 1 items
Catalog # METB30029066
Supplier:  Mettler - Toledo
Frequently Bought Together