Easy-fit torch, demountable for semi-volatile organics, with replaceable 1.4 mm id injector and base assembly, with replaceable oute r and intermediate tubes, for use with Agilent 5100 Synchronous Vertical Dual View/Vertical Dual View ICP-OES in axia 1 * 1 items
Catalog # HEWLG8010-60233
Supplier: Agilent
Frequently Bought Together