REPLIGEN CORP. RSPA NATIVE RECOMBINANT S. AUREUS PROTEIN A (ANIMAL FREE) 1 * 10 mg
Catalog # MSPP10-2001-XM
Supplier: MARKET SOURCE PART PROCESS
REPLIGEN CORP. RSPA NATIVE RECOMBINANT S. AUREUS PROTEIN A (ANIMAL FREE) 1 * 10 mg
Catalog # MSPP10-2001-XM
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection