Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
COVERALL COLOR: WHITE SIZE: XL DESCRIPTION: ZIPPER IN FRONT, WITH HOOD, LATEX-FREE ELASTIC ON WRIST/ANKLE/WAIST MATERIAL: POLYPROPYLENE THICKNESS: 40 G/M PACKAGING: 1 PC/BAG, 50 PCS/CARTON 1 * 50 items
GUNA140034
:
VWR Collection
COVERALL COLOR: WHITE SIZE: XL DESCRIPTION: ZIPPER IN FRONT, WITH HOOD, LATEX-FREE ELASTIC ON WRIST/ANKLE/WAIST MATERIAL: POLYPROPYLENE THICKNESS: 40 G/M PACKAGING: 1 PC/BAG, 50 PCS/CARTON 1 * 50 items
GUNA140034
:
VWR Collection
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection