Cortisol ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cortisol in dried fecal extracts, saliva, urine, serum, plasma and tissue culture media samples 1 * 1 KIT
Catalog # ABNOKA3382
Supplier: Abnova
Frequently Bought Together