ContactUsLinkComponent
Capillary kit, for Agilent Online-SPE Starter Set G4742A. Includes 5 capillaries, stainless steel, 0.12 x 120 mm, pre-swaged (5067-4 652), 2 capillaries, stainless steel, 0.12 x 340 mm, S/SX fittings (5067-4647), capillary, stainless steel, 0.25 x 50 1 * 1 items
Catalog # HEWL5067-5708
Supplier:  Agilent
Capillary kit, for Agilent Online-SPE Starter Set G4742A. Includes 5 capillaries, stainless steel, 0.12 x 120 mm, pre-swaged (5067-4 652), 2 capillaries, stainless steel, 0.12 x 340 mm, S/SX fittings (5067-4647), capillary, stainless steel, 0.25 x 50 1 * 1 items
Catalog # HEWL5067-5708
Supplier:  Agilent
Frequently Bought Together