ContactUsLinkComponent
Quartz glass capsule in application temperatures of up to 1050°C (maximum 1600°C), round, translucent, high chemical resistance and purity. Diameter 120mm, height 120mm. 1 * 1 items
Catalog # TSLA32K1203
Quartz glass capsule in application temperatures of up to 1050°C (maximum 1600°C), round, translucent, high chemical resistance and purity. Diameter 120mm, height 120mm. 1 * 1 items
Catalog # TSLA32K1203
Frequently Bought Together