ContactUsLinkComponent
Chromosorb PNAW 1.2% Dc-200/500, 80/100, preconditioned, 20 inch (0.51 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81177
Supplier:  Agilent
Chromosorb PNAW 1.2% Dc-200/500, 80/100, preconditioned, 20 inch (0.51 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81177
Supplier:  Agilent
Frequently Bought Together