ContactUsLinkComponent
Chromosorb PAW 25% Dc-200, 80/100, preconditioned, 41.28 mm coil, 15 feet (4.57 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81201
Supplier:  Agilent
Chromosorb PAW 25% Dc-200, 80/100, preconditioned, 41.28 mm coil, 15 feet (4.57 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81201
Supplier:  Agilent
Frequently Bought Together