Orders LinkContactUsLinkComponent
Clino Mopping System
Clino Mopping System
Clino mop holder, magnet, Grey, 400 mm
Catalog # PPSA2500050
Clino Mopping System
Catalog # PPSA2500050
Clino Mopping System
Clino Mopping System
Clino mop holder, magnet, Grey, 400 mm
Price
43,16 €
Log in for availability
1 * 1 items
Product Details & Documents
Mop frame is suitable for use with all Pfennig mop heads: standard, professional, and universal.
  • Sturdy and durable stainless steel construction
  • Sterilizable
  • Contact-free mop drop
  • Also available with locking-screw
 : Mop frames and handles are sold separately. Telescopic alumimium mop handle must be ordered along with a mop frame to complete the system.
Specifications
  • Colour:
    Grey
  • Description:
    Clino mop holder, magnet
  • Length:
    400 mm
  • Seq:
    04