ContactUsLinkComponent
Chromaster HPLC Instrument Package with 5160 600 Bar Manual Purge Pump, LPG, Degasser, UI Pad, Standard Autosampler, Column Oven, UV Detector, Organiser, Dell PC with Win7 32 Bit operating system, keyboard mouse and monitor and Chromaster System Mana 1 * 1 items
Catalog # 903-0300
Supplier:  VWR Collection
Chromaster HPLC Instrument Package with 5160 600 Bar Manual Purge Pump, LPG, Degasser, UI Pad, Standard Autosampler, Column Oven, UV Detector, Organiser, Dell PC with Win7 32 Bit operating system, keyboard mouse and monitor and Chromaster System Mana 1 * 1 items
Catalog # 903-0300
Supplier:  VWR Collection
Frequently Bought Together