qPCR detection of Coxiella burnetii. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome virus es only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.BUR
Supplier: PRIMERDESIGN
Frequently Bought Together