qPCR detection of Human Polyomavirus 6. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vi ruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HPYV6
Supplier: PRIMERDESIGN
Frequently Bought Together