ContactUsLinkComponent
qPCR detection of Porcine Reproductive and Respiratory Syndrome Virus. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
  PATH-PRRSV
qPCR detection of Porcine Reproductive and Respiratory Syndrome Virus. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
  PATH-PRRSV
Frequently Bought Together