ContactUsLinkComponent
MOBCAP COLOR: YELLOW SIZE: 52CM DESCRIPTION: DETECTABLE WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 12-14 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 items
Catalog # GUNA140016
Supplier:  VWR Collection
MOBCAP COLOR: YELLOW SIZE: 52CM DESCRIPTION: DETECTABLE WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 12-14 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 items
Catalog # GUNA140016
Supplier:  VWR Collection
Frequently Bought Together