ContactUsLinkComponent
Quantitative PCR (qPCR also known as real time PCR) simultaneously amplifies and detects targeted DNA allowing scientists to discove r the starting amount of a specific DNA sequence in an experimental sample. 1 * 1 KIT
Catalog # EDVO380
Supplier:  EDVOTEK
Quantitative PCR (qPCR also known as real time PCR) simultaneously amplifies and detects targeted DNA allowing scientists to discove r the starting amount of a specific DNA sequence in an experimental sample. 1 * 1 KIT
Catalog # EDVO380
Supplier:  EDVOTEK
Frequently Bought Together