Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
1.000 mL
Catalog # J63202.AU
Flammable
Health
Irritant
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
Catalog # J63202.AU
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use
1.000 mL
CAS Number:  54827-17-7
Price
500,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
  • ADR:3,II
Specifications
Specifications
  • Pk:
    1.000 mL