ContactUsLinkComponent
SAA1 monkey Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 104 amino acids and having to tal molecular mass of 11.8 kDa. SAA1 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80101-2
Supplier:  Biorbyt
SAA1 monkey Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 104 amino acids and having to tal molecular mass of 11.8 kDa. SAA1 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80101-2
Supplier:  Biorbyt