ContactUsLinkComponent
IL15 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 114 amino acids and having a mo lecular mass of 12774 Dalton. The IL-15 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80076-2
Supplier:  Biorbyt
IL15 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 114 amino acids and having a mo lecular mass of 12774 Dalton. The IL-15 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80076-2
Supplier:  Biorbyt