Interferon-gamma Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 144 amino acids an d having a molecular mass of 16879 Dalton.The IFN-gamma is purified by proprietary chromatographic technique 1 * 100 µG
Catalog # BIRBORB80041-100
Supplier: Biorbyt
Frequently Bought Together