ContactUsLinkComponent
FGF19 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 195 amino acids and having a molecular mass of 21.8 kDa.The FGF-19 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80027-5
Supplier:  Biorbyt
FGF19 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 195 amino acids and having a molecular mass of 21.8 kDa.The FGF-19 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80027-5
Supplier:  Biorbyt
Related Information
Frequently Bought Together