MICA Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide containing 320 amino acids and having a molecul ar mass of 36kDa. The sequence contains the full length extracellular domain of the mature human MICA (amino 1 * 10 µG
Catalog # BIRBORB80176-10
Supplier: Biorbyt
Frequently Bought Together