CNTF Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a mo lecular mass of 22706 Dalton. The CNTF is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80012-20
Supplier: Biorbyt
Frequently Bought Together