ContactUsLinkComponent
Beta Defensin-2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 41 amino acids and h aving a molecular mass of 4.3 kDa. The BD-2 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80002-20
Supplier:  Biorbyt
Beta Defensin-2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 41 amino acids and h aving a molecular mass of 4.3 kDa. The BD-2 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80002-20
Supplier:  Biorbyt