ContactUsLinkComponent
HTLV-1 Envelope protein.The protein contains the C-terminus of gp46 and mostof p21E of HTLV-1. This non-fusion E.coli derived protei nstarts from HIV-1 envelope amino acids 165 and ends with440.The protein is purified by proprietary chromato 1 * 1 mg
Catalog # BIRBORB85277-1
Supplier:  Biorbyt
HTLV-1 Envelope protein.The protein contains the C-terminus of gp46 and mostof p21E of HTLV-1. This non-fusion E.coli derived protei nstarts from HIV-1 envelope amino acids 165 and ends with440.The protein is purified by proprietary chromato 1 * 1 mg
Catalog # BIRBORB85277-1
Supplier:  Biorbyt
Related Information
Frequently Bought Together