ContactUsLinkComponent
Enterokinase Bovine Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 241 amino acids with a 6 multiply His at C-terminus and having a molecular mass of 28.0kDa. 1 * 1.000 EU
Catalog # BIRBORB82608-1000
Supplier:  Biorbyt
Enterokinase Bovine Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 241 amino acids with a 6 multiply His at C-terminus and having a molecular mass of 28.0kDa. 1 * 1.000 EU
Catalog # BIRBORB82608-1000
Supplier:  Biorbyt
Related Information
Frequently Bought Together