ContactUsLinkComponent
B2 Microglobulin Human Recombinant produced in E.coli is non-glycosylated polypeptide chain having a molecular mass of 11.76 kDa.The B2M is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80944-10
 :  Biorbyt
B2 Microglobulin Human Recombinant produced in E.coli is non-glycosylated polypeptide chain having a molecular mass of 11.76 kDa.The B2M is purified by proprietary chromatographic techniques. 1 * 10 µG
  BIRBORB80944-10
 :  Biorbyt
Frequently Bought Together