Human Myoglobin produced in Human Cardiac Tissues having a molecular mass of 17.5kDa. Myoglobin is released from recently injured my ocardial cells within few hours of Infarction. Peak levels are reached more quickly than CK-MB or Troponin c 1 * 200 µG
BIRBORB81227-200
: Biorbyt
Frequently Bought Together