ContactUsLinkComponent
YWHAZ fused to His Tag on N-terminus Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containin g 245amino acids (1-245) and having a molecular mass of 32 kDa. YWHAZ is purified by proprietary chromatogra 1 * 5 µG
Catalog # BIRBORB80886-5
Supplier:  Biorbyt
YWHAZ fused to His Tag on N-terminus Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containin g 245amino acids (1-245) and having a molecular mass of 32 kDa. YWHAZ is purified by proprietary chromatogra 1 * 5 µG
Catalog # BIRBORB80886-5
Supplier:  Biorbyt