ContactUsLinkComponent
LBP is produced from mouse LBP transfected CHO-cells in serum free medium. For transfection we have cloned complete mouse LBP cDNA i nto expression vector pPOL-DHFR. Before tranfection the complete mouse LBP cDNA was amplified by PCR and clo 1 * 5 µG
Catalog # BIRBORB81195-5
Supplier:  Biorbyt
LBP is produced from mouse LBP transfected CHO-cells in serum free medium. For transfection we have cloned complete mouse LBP cDNA i nto expression vector pPOL-DHFR. Before tranfection the complete mouse LBP cDNA was amplified by PCR and clo 1 * 5 µG
Catalog # BIRBORB81195-5
Supplier:  Biorbyt