Recombinant Full-Length H9N2 A/Hong-Kong/1073/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is 72,000 dalton. The accession number is AJ404626. 1 * 2 µG
BIRBORB81785-2
: Biorbyt
Frequently Bought Together