Recombinant Full-Length H3N2 A/Wisconsin/67/05 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect ce lls and its Mw is 70,000 dalton. 1 * 2 µG
Recombinant Full-Length H3N2 A/Wisconsin/67/05 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect ce lls and its Mw is 70,000 dalton. 1 * 2 µG