Cystatin C or cystatin, a protein encoded by the CST3 gene. Cystatin 3 has a low molecular weight (approximately 13.3kDa), and is re adily removed from the bloodstream by glomerular filtration in the kidneys, the rise of cystatin c in the bl 1 * 96 items
Catalog # BIRBORB50140-96
Supplier: Biorbyt
Frequently Bought Together