Recombinant Human TNC produced in E.coli is a single, non-glycosylated, polypeptide chain having a molecular mass of 18,020 Dalton.T he TNC is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81436-20
Supplier: Biorbyt
Frequently Bought Together