ContactUsLinkComponent
Recombinant Human THBS1 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. Recombinant Hum an THBS1 is Reactive with A4.1 anti-TSP mAb and its Mw is 140,000 Dalton. 1 * 2 µG
Catalog # BIRBORB81417-2
Supplier:  Biorbyt
Recombinant Human THBS1 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. Recombinant Hum an THBS1 is Reactive with A4.1 anti-TSP mAb and its Mw is 140,000 Dalton. 1 * 2 µG
Catalog # BIRBORB81417-2
Supplier:  Biorbyt
Related Information
Frequently Bought Together